missing translation for 'onlineSavingsMsg'
Learn More

Methyl-alpha-D-mannopyranoside, 99%

Product Code. 11474543
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474543 100 g 100g
11464543 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474543 Supplier Thermo Scientific Alfa Aesar Supplier No. A11533.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.

Solubility
It is easily soluble in methanol, water. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agent. Stable under recommended storage conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 193°C to 196°C
Quantity 100 g
Beilstein 81566
Solubility Information It is easily soluble in methanol,water. Insoluble in ether.
Optical Rotation +79° (c=10 in water)
Formula Weight 194.18
Percent Purity 99%
Chemical Name or Material Methyl-alpha-D-mannopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.