missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-chloroacrylate, 98+%, stabilized with hydroquinone

Product Code. 10771632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10771632 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10771632 Supplier Thermo Scientific Acros Supplier No. 414160250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-63-7 | C4H5ClO2 | 120.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80-63-7
Color Colorless to Brown
Density 1.1890g/mL
Boiling Point 52°C (50.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5ClO2
Linear Formula CH2CClCOOCH3
Refractive Index 1.4420 to 1.444
MDL Number MFCD00051368
Quantity 25 g
Specific Gravity 1.189
Synonym methyl 2-chloroacrylate, methyl alpha-chloroacrylate, methyl-2-chloroacrylate, 2-propenoic acid, 2-chloro-, methyl ester, methyl-alpha-chloroacrylate, 2-chloroacrylic acid, methyl ester, methyl 2-chloro-2-propenoate, acrylic acid, 2-chloro-, methyl ester, methyl 2-chloro-2-propenate, unii-47pg4l077j
Solubility Information Solubility in water: .. Other solubilities: soluble in diethylether
InChI Key AWJZTPWDQYFQPQ-UHFFFAOYSA-N
SMILES COC(=O)C(=C)Cl
IUPAC Name methyl 2-chloroprop-2-enoate
Stabilizer abt. 0.06% hydroquinone
Molecular Weight (g/mol) 120.54
PubChem CID 6659
Formula Weight 120.54
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl α-chloroacrylate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl a-chloroacrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.