missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-bromophenylacetate, 97%, Thermo Scientific™

Product Code. 11372846
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372846 5 g 5g
11382846 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372846 Supplier Thermo Scientific Alfa Aesar Supplier No. L09043.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3042-81-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00013535
InChI Key NHFBYYMNJUMVOT-UHFFFAOYSA-N
Synonym methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate
PubChem CID 137806
IUPAC Name methyl 2-bromo-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)Br

Specifications

Boiling Point 90°C to 92°C (0.5 mmHg)
Flash Point 150°C (302°F)
Quantity 5 g
UN Number UN3265
Beilstein 1102743
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl alpha-bromophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.