missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-Alpha,Alpha-diphenyl-4-morpholinebutyronitrile(Regioisomer Mixture), TRC
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
Specifications
Specifications
| Chemical Name or Material | Methyl-alpha,alpha-diphenyl-4-morpholinebutyronitrile(Regioisomer Mixture) |
| Molecular Formula | C21H24N2O |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/2C21H24N2O/c1-18(16-23-12-14-24-15-13-23)21(17-22,19-8-4-2-5-9-19)20-10-6-3-7-11-20;1-18(23-12-14-24-15-13-23)16-21(17-22,19-8-4-2-5-9-19)20-10-6-3-7-11-20/h2*2-11,18H,12-16H2,1H3 |
| SMILES | CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)C#N)CN3CCOCC3.CC(CC(C4=CC=CC=C4)(C5=CC=CC=C5)C#N)N6CCOCC6 |
| Molecular Weight (g/mol) | 320.43 |
| Formula Weight | 320.19 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction