missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Alcohol, NF, Macron Fine Chemicals™
Click to view available options
Quantity:
4 L
Packaging:
SAFEMOR PVC-coated Glass Bottle
Unit Size:
4 L
Description
- Meets the requirements necessary for use in pharmaceutical processing and prescription compounding
- Product is regulated by the Food and Drug Administration
- Manufactured under current Good Manufacturing Practices (cGMP)
Specifications
Specifications
| Melting Point | -97.6°C |
| CAS | 67-56-1 |
| Color | Colorless |
| Acidity | Passes test |
| Alkalinity | ≤3 ppm (as NH3) |
| Appearance | Clear |
| Density | 0.80g/mL |
| Boiling Point | 64.7°C |
| Quantity | 4 L |
| Flash Point | 12.0°C |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H361-Suspected of damaging fertility or the unborn child.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction