missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetopyruvate, 98%, Thermo Scientific™

Product Code. 11443494
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443494 Supplier Thermo Scientific Alfa Aesar Supplier No. A11190.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20577-61-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00043638
InChI Key OMHOEQINEXASKE-UHFFFAOYSA-N
Synonym methyl acetopyruvate, methyl acetylpyruvate, pentanoic acid, 2,4-dioxo-, methyl ester, 2,4-dioxopentanoic acid methyl, 2,4-dioxo-pentanoic acid methyl ester, pentanoic acid,2,4-dioxo-,methyl ester
PubChem CID 88600
IUPAC Name methyl 2,4-dioxopentanoate
SMILES CC(=O)CC(=O)C(=O)OC

Specifications

Melting Point 60°C to 63°C
Quantity 25 g
Formula Weight 144.13
Percent Purity 98%
Chemical Name or Material Methyl acetopyruvate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.