missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetoacetate, Mikromol™

Product Code. 30050227
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050227 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050227 Supplier Mikromol Supplier No. MM0003.100100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl Acetoacetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetoacetic acid methyl ester
CAS 105-45-3
Analyte or Component Names Acetoacetic acid methyl ester
Shipping Condition Room Temperature
Molecular Formula C5 H8 O3
Certifications/Compliance ISO 9001
Synonym Butanoic acid, 3-oxo-, methyl ester, Acetoacetic acid, methyl ester (6CI,8CI), 3-Oxobutanoic acid methyl ester, 3-Oxobutyric acid methyl ester, Acetoacetate methyl ester, Methyl 3-oxobutanoate, Methyl 3-oxobutyrate, Methyl acetoacetate, Methyl acetylacetate
InChI Formula InChI=1S/C5H8O3/c1-4(6)3-5(7)8-2/h3H2,1-2H3
SMILES COC(=O)CC(=O)C
IUPAC Name methyl 3-oxobutanoate
Molecular Weight (g/mol) 116.12
Quantity 100 mg
Formula Weight 116.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.