missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate-d6, TRC

Product Code. 30484238
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484238 50 mg 50mg
30480247 10 mg 10mg
30474329 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30484238 Supplier Toronto Research Chemicals Supplier No. TRCM27535250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32113-85-2
Molecular Formula C3 D6 O2
Molecular Weight (g/mol) 80.12
Synonym Acetic-d3 acid, methyl-d3 ester (6CI, 8CI, 9CI), Methyl Acetate-d6, Methyl Acetate-D6, Acetic-d3 acid methyl-d3 ester, Acetic acid methyl ester-d6
IUPAC Name trideuteriomethyl
SMILES [2H]C([2H])([2H])OC(=O)C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H6O2/c1-3(4)5-2/h1-2H3/i1D3,2D3
Formula Weight 80.0744
Chemical Name or Material Methyl Acetate-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.