missing translation for 'onlineSavingsMsg'
Learn More

Methyl 9-octadecynoate, 90+%, Thermo Scientific™

Product Code. 15413367
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413367 Supplier Thermo Scientific Alfa Aesar Supplier No. H26080.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-32-7
Molecular Formula C19H34O2
MDL Number MFCD09953468
Quantity 1 g
Synonym methyl stearolate, methyl 9-octadecynoate, 9-octadecynoic acid methyl ester, 9-octadecynoic acid, methyl ester, methyl 9-octadecynoate #, acmc-1br0q, stearolic acid methyl ester, methyl 9-octadecynoate, 90+%, 9-octadecynoic acid,methyl ester
InChI Key VGXHOYHYUMFLPG-UHFFFAOYSA-N
SMILES CCCCCCCCC#CCCCCCCCC(=O)OC
IUPAC Name methyl octadec-9-ynoate
Molecular Weight (g/mol) 294.479
PubChem CID 534587
Formula Weight 294.47
Percent Purity ≥90%
Chemical Name or Material Methyl 9-octadecynoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.