missing translation for 'onlineSavingsMsg'
Learn More

Methyl 9,10-Dihydroxystearate, TRC

Product Code. 30476792
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476792 1 g 1g
30467503 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476792 Supplier Toronto Research Chemicals Supplier No. TRCM3027501G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-01-1
Molecular Formula C19H38O4
Molecular Weight (g/mol) 330.5
Synonym 9,10-Dihydroxystearic Acid Methyl Ester, Methyl 9,10-Dihydroxyoctadecanoate, Methyl 9,10-Dihydroxystearate, NSC 409398
IUPAC Name methyl 9,10-dihydroxyoctadecanoate
SMILES OC(CCCCCCCC(OC)=O)C(CCCCCCCC)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H38O4/c1-3-4-5-6-8-11-14-17(20)18(21)15-12-9-7-10-13-16-19(22)23-2/h17-18,20-21H,3-16H2,1-2H3
Formula Weight 330.28
Chemical Name or Material Methyl 9,10-Dihydroxystearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.