missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-phenylheptanoate, 98%

Product Code. 11324825
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324825 5 g 5g
11314825 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324825 Supplier Thermo Scientific Alfa Aesar Supplier No. L17905.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101689-18-3 | C14H20O2 | 220.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101689-18-3
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.31
MDL Number MFCD02093971
InChI Key FZSNATXOGRAOIL-UHFFFAOYSA-N
Synonym acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester
PubChem CID 7010318
IUPAC Name methyl 7-phenylheptanoate
SMILES COC(=O)CCCCCCC1=CC=CC=C1

Specifications

Boiling Point ∼155°C (1.5mmHg)
Refractive Index 1.4946
Quantity 5 g
Beilstein 2694776
Formula Weight 220.31
Percent Purity 98%
Chemical Name or Material Methyl 7-phenylheptanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.