missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-bromo-3-chloro-4-fluorobenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. p-7052545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419075 1 g 1g
15429075 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419075 Supplier Thermo Scientific Alfa Aesar Supplier No. H36325.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1823304-13-7
Molecular Formula C10H5BrClFO2S
Molecular Weight (g/mol) 323.56
MDL Number MFCD23380840
InChI Key UHFJFXCPHLTILG-UHFFFAOYSA-N
Synonym 7-Bromo-3-chloro-4-fluorobenzo[b]thiophene-2-carboxylic acidmethyl ester
PubChem CID 98001137
IUPAC Name methyl 7-bromo-3-chloro-4-fluoro-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C2=C(C=CC(=C2S1)Br)F)Cl

Specifications

Melting Point 149°C to 151°C
Color Yellow
Quantity 1 g
Formula Weight 323.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 7-bromo-3-chloro-4-fluorobenzo[b]thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.