missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Oxohexanoate, TRC

Product Code. 30472679
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472679 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472679 Supplier Toronto Research Chemicals Supplier No. TRCM3258501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 6-Oxohexanoate
CAS 6654-36-0
Molecular Formula C7 H12 O3
Quantity 1 g
Synonym Hexanoic acid, 6-oxo-, methyl ester, Adipaldehydic acid, methyl ester (6CI,7CI,8CI), 5-Carbomethoxy-1-pentanal, 5-Formylpentanoic acid, methyl ester, 5-Formylvaleric acid, methyl ester, 6-Oxohexanoic acid methyl ester, Adipic acid semialdehyde methyl ester, Methyl 5-formylpentanoate, Methyl 5-formylvalerate, Methyl 6-oxocaproate, Methyl 6-oxohexanoate, Methyl adipaldehydate
InChI Formula InChI=1S/C7H12O3/c1-10-7(9)5-3-2-4-6-8/h6H,2-5H2,1H3
SMILES COC(=O)CCCCC=O
IUPAC Name methyl 6-oxohexanoate
Molecular Weight (g/mol) 144.17
Formula Weight 144.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.