missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-methylnicotinate, 97+%

Product Code. 11398715
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398715 5 g 5g
11388715 1 g 1g
11308725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398715 Supplier Thermo Scientific Alfa Aesar Supplier No. L12276.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5470-70-2 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-70-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006340
InChI Key VYPPZXZHYDSBSJ-UHFFFAOYSA-N
Synonym methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
PubChem CID 231548
IUPAC Name methyl 6-methylpyridine-3-carboxylate
SMILES CC1=NC=C(C=C1)C(=O)OC

Specifications

Melting Point 31°C to 36°C
Boiling Point 152°C to 154°C (40 mmHg)
Flash Point 103°C (217°F)
Quantity 5 g
Beilstein 122940
Formula Weight 151.17
Percent Purity ≥97%
Chemical Name or Material Methyl 6-methylnicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.