missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloropyridazine-4-carboxylate, 97%

Product Code. 10098289 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10098289 1 g Glass bottle 1g
10331230 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10098289 Supplier Thermo Scientific Acros Supplier No. 432980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1093860-48-0 | C6H5ClN2O2 | 172.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093860-48-0
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD16660968
InChI Key QYQHGLKSLGUINZ-UHFFFAOYSA-N
Synonym 6-chloro-pyridazine-4-carboxylic acid methyl ester, methyl6-chloropyridazine-4-carboxylate, 4-pyridazinecarboxylicacid, 6-chloro-, methyl ester, 6-chloro-4-pyridazinecarboxylic acid, methyl ester, 4-pyridazinecarboxylic acid, 6-chloro-, methyl ester
PubChem CID 45790080
IUPAC Name methyl 6-chloropyridazine-4-carboxylate
SMILES COC(=O)C1=CC(Cl)=NN=C1

Specifications

Melting Point 88.4°C to 88.5°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl 6-chloropyridazine-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.