missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Chloronicotinate, TRC

Product Code. 30475506
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475506 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475506 Supplier Toronto Research Chemicals Supplier No. TRCM29527525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 6-Chloronicotinate
CAS 73781-91-6
Molecular Formula C7H6ClNO2
Quantity 25 g
Synonym 6-Chloro-3-pyridinecarboxylic Acid Methyl Ester, 2-Chloro-5-(methoxycarbonyl)pyridine, 6-Chloronicotinic Acid Methyl Ester, 6-Chloropyridine-3-carboxylic Acid Methyl Ester, Methyl 2-Chloropyridin-5-ylcarboxylate, Methyl 2-Chloropyridine-5-carboxylate, Methyl 6-Chloro-3-pyridinecarboxylate, NSC 78316
InChI Formula InChI=1S/C7H6ClNO2/c1-11-7(10)5-2-3-6(8)9-4-5/h2-4H,1H3
SMILES COC(=O)c1ccc(Cl)nc1
IUPAC Name methyl 6-chloropyridine-3-carboxylate
Molecular Weight (g/mol) 171.58
Formula Weight 171.0087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.