missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloronicotinate, 98%

Product Code. 11418583
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418583 1 g 1g
11428583 5 g 5g
11438583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418583 Supplier Thermo Scientific Alfa Aesar Supplier No. A10396.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 73781-91-6 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 73781-91-6
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00023420
InChI Key RMEDXVIWDFLGES-UHFFFAOYSA-N
PubChem CID 254249
IUPAC Name methyl 6-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)Cl

Especificaciones

Melting Point 83°C to 88°C
Quantity 1 g
Beilstein 128648
Synonym methyl 6-chloronicotinate, 6-chloronicotinic acid methyl ester, methyl6-chloronicotinate, methyl 2-chloropyridine-5-carboxylate, 3-pyridinecarboxylic acid, 6-chloro-, methyl ester, methyl 6-chloro-3-pyridinecarboxylate, 6-chloro-3-pyridinecarboxylic acid methyl ester, pubchem13584, methyl-6-chloronicotinate, methyl 6-chloro-nicotinate
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material Methyl 6-chloronicotinate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.