missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromo-5-fluoronicotinate, 98%

Product Code. 15441589
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441589 5 g 5g
15451589 250 mg 250mg
15431589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441589 Supplier Thermo Scientific Alfa Aesar Supplier No. H64395.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1214336-88-5 | C7H5BrFNO2 | 234.02 g/mol

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent. Store in freezer at (-20°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214336-88-5
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.02
MDL Number MFCD14698110
InChI Key PXHKAVXJQLVUBI-UHFFFAOYSA-N
Synonym methyl 6-bromo-5-fluoronicotinate, 6-bromo-5-fluoro-nicotinic acid methyl ester, 2-bromo-3-fluoro-5-methoxycarbonyl pyridine, methyl6-bromo-5-fluoronicotinate, abbypharma ap-30-7441, 6-bromo-5-fluoropyridine-3-carboxylate, methyl 2-bromo-3-fluoro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-bromo-5-fluoro-, methyl ester
PubChem CID 46311203
IUPAC Name methyl 6-bromo-5-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CC(F)=C(Br)N=C1

Specifications

Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 234.02
Percent Purity 98%
Chemical Name or Material Methyl 6-bromo-5-fluoronicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.