missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromo-2-naphthoate, 98%

Product Code. 11456137
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456137 25 g 25g
11436137 1 g 1g
11446137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456137 Supplier Thermo Scientific Alfa Aesar Supplier No. B25106.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33626-98-1 | C12H9BrO2 | 265.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33626-98-1
Molecular Formula C12H9BrO2
Molecular Weight (g/mol) 265.106
MDL Number MFCD00100408
InChI Key JEUBRLPXJZOGPX-UHFFFAOYSA-N
Synonym methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate
PubChem CID 854134
IUPAC Name methyl 6-bromonaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 121°C to 123°C
Quantity 25 g
Beilstein 2578943
Formula Weight 265.11
Percent Purity 98%
Chemical Name or Material Methyl 6-bromo-2-naphthoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.