missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-aminonicotinate, 97%

Product Code. 11416470
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416470 1 g 1g
11426470 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416470 Supplier Thermo Scientific Alfa Aesar Supplier No. H27575.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36052-24-1 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36052-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00797844
InChI Key JACPDLJUQLKABC-UHFFFAOYSA-N
Synonym methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
PubChem CID 737487
IUPAC Name methyl 6-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N

Specifications

Melting Point 158°C to 161°C
Quantity 1 g
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Methyl 6-aminonicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.