missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-aminoindole-4-carboxylate hydrochloride, 97%, Thermo Scientific™

Product Code. 15428965
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 15428965

Brand: Thermo Scientific Alfa Aesar H34475.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 6-aminoindole-4-carboxylate hydrochloride is used as an organic chemical synthesis intermediate.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 731810-08-5
Melting Point >230°C
Molecular Formula C10H11ClN2O2
MDL Number MFCD09258897
Quantity 500 mg
Sensitivity Hygroscopic
Synonym methyl 6-amino-1h-indole-4-carboxylate hydrochloride, methyl 6-aminoindole-4-carboxylate hydrochloride, 1h-indole-4-carboxylic acid, 6-amino-, methyl ester, monohydrochloride, 1h-indole-4-carboxylicacid, 6-amino-, methyl ester, hydrochloride 1:1, 6-aminoindole-4-carboxylic acid methyl ester hydrochloride, 6-amino-1h-indole4-carboxylic acid methyl ester hydrochloride
InChI Key QMSZHGOALNOGQC-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(=CC2=C1C=CN2)N.Cl
IUPAC Name methyl 6-amino-1H-indole-4-carboxylate;hydrochloride
Molecular Weight (g/mol) 226.66
PubChem CID 46741374
Formula Weight 226.66
Percent Purity 97%
Chemical Name or Material Methyl 6-aminoindole-4-carboxylate hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.