missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-amino-5-bromonicotinate, 98%, Thermo Scientific™

Product Code. 10585860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10585860 5 g Glass bottle 5g
10351380 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10585860 Supplier Thermo Scientific Acros Supplier No. 437940050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 180340-70-9
Molecular Formula C7H8BrN2O2
Molecular Weight (g/mol) 232.06
MDL Number MFCD06659561
InChI Key LNTAAMVZKGYIIA-UHFFFAOYSA-O
Synonym methyl 6-amino-5-bromonicotinate, methyl 2-amino-3-bromo-5-pyridinecarboxylate, methyl 2-amino-3-bromopyridine-5-carboxylate, methyl-6-amino-5-bromonicotinate, 3-pyridinecarboxylic acid, 6-amino-5-bromo-, methyl ester, 6-amino-5-bromonicotinic acid methyl ester, pubchem9472, acmc-209egj, methyl 6-amino-5-bromo-pyridine-3-carboxylate, methyl-6-5-bromonicotinate
PubChem CID 2764099
IUPAC Name methyl 6-amino-5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=C[NH+]=C(N)C(Br)=C1

Specifications

Melting Point 173.5°C to 175.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 231.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 6-amino-5-bromonicotinate
compliance-icons
Product Identifier
  • Methyl 6-amino-5-bromonicotinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.