missing translation for 'onlineSavingsMsg'
Learn More

Methyl [5-Propylsulphonyl)-1H-benzimidazol-2-yl]carbamate, Mikromol™

Product Code. 17931796
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931796 100 mg 100mg
17952656 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17931796 Supplier Mikromol Supplier No. MM0382.03
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl [5-Propylsulphonyl)-1H-benzimidazol-2-yl]carbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Albendazole sulfone
CAS 75184-71-3
Analyte or Component Names Albendazole sulfone
Shipping Condition Room Temperature
Molecular Formula C12 H15 N3 O4 S
Certifications/Compliance ISO 17025
Synonym Carbamic acid, [5-(propylsulfonyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Methyl [5-(propylsulfonyl)-1H-benzimidazol-2-yl]carbamate, Methyl [5-Propylsulphonyl)-1H-benzimidazol-2-yl]carbamate, Albendazole Imp. C (EP), SKF-63896, Ph Eur Albendazole Impurity C, Albendazole Impurity C
InChI Formula InChI=1S/C12H15N3O4S/c1-3-6-20(17,18)8-4-5-9-10(7-8)14-11(13-9)15-12(16)19-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
SMILES CCCS(=O)(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1
IUPAC Name methyl N-(5-propylsulfonyl-1H-benzimidazol-2-yl)carbamate
Molecular Weight (g/mol) 297.33
Quantity 100 mg
Formula Weight 297.0783 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.