missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech.

Product Code. 11434433
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11434433

Brand: Thermo Scientific Alfa Aesar A10982.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25134-21-8 | C10H10O3 | 178.19 g/mol

Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

Solubility
Miscible with acetone, benzene, naphtha and xylene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25134-21-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00167036
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym Methyl nadic anhydride

Specifications

Density 1.232
Boiling Point 137°C to 140°C (10 mmHg)
Flash Point 135°C (275°F)
Refractive Index 1.5052
Quantity 500 g
Sensitivity Moisture sensitive
Solubility Information Miscible with acetone,benzene,naphtha and xylene.
Formula Weight 178.19
Grade Technical
Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.