missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers

Product Code. 10082830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082830 1 kg Glass bottle 1kg
10472791 250 g Glass bottle 250g
10513514 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082830 Supplier Thermo Scientific Acros Supplier No. 216270010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25134-21-8 | C10H10O3 | 178.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25134-21-8
MDL Number MFCD00167036
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride

Specifications

Color Yellow
Density 1.2300g/mL
Boiling Point 275.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C10H10O3
Refractive Index 1.5060 to 1.5080
Quantity 1 kg
Beilstein 17, 461
Specific Gravity 1.23
Molecular Weight (g/mol) 178.19
Formula Weight 178.18
Percent Purity 95%
Physical Form Oily Liquid
Chemical Name or Material Methyl-5-norbornene-2, 3-dicarboxylic anhydride, Mixture of isomers
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.