missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylnicotinate, 94%

Product Code. 11389135
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389135 5 g 5g
11379135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389135 Supplier Thermo Scientific Alfa Aesar Supplier No. L16371.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29681-45-6 | C8H9NO2 | 151.165 g/mol

5-Methyl-N?-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N?-nitrosonornicotine is used as the starting material in organic compound synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-N′-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N′-nitrosonornicotine is used as the starting material in organic compound synthesis.

Solubility
Soluble in Chloroform and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-45-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD01075739
InChI Key KQILMMLAGGFMCM-UHFFFAOYSA-N
PubChem CID 256308
IUPAC Name methyl 5-methylpyridine-3-carboxylate
SMILES CC1=CN=CC(=C1)C(=O)OC

Specifications

Melting Point 42°C to 46°C
Boiling Point 90°C (3 mmHg)
Quantity 5 g
Beilstein 472498
Synonym methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej
Solubility Information Soluble in Chloroform and Methanol.
Formula Weight 151.17
Percent Purity 94%
Chemical Name or Material Methyl 5-methylnicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.