missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylisoxazole-3-carboxylate, 97%

Product Code. 11385279
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385279 25 g 25g
11395279 100 g 100g
11305289 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385279 Supplier Thermo Scientific Alfa Aesar Supplier No. A19814.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19788-35-3 | C6H7NO3 | 141.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19788-35-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD00015895
InChI Key MVHHQOCEOUNTID-UHFFFAOYSA-N
Synonym methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
PubChem CID 88245
IUPAC Name methyl 5-methyl-1,2-oxazole-3-carboxylate
SMILES COC(=O)C1=NOC(C)=C1

Specifications

Melting Point 91°C to 92°C
Boiling Point 125°C (11 mmHg)
Quantity 25 g
Beilstein 115704
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material Methyl 5-methylisoxazole-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.