missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methyl-2-furoate, 97%

Product Code. 15463717
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463717 Supplier Thermo Scientific Alfa Aesar Supplier No. H27570.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2527-96-0 | C7H8O3 | 140.138 g/mol

Furans with suitable substituent in the 2- positions may be transformed into other systems with aromatic character with methoxylation of cyclic acetals of 1,4- dicarbonyl compounds, followed by intramolecular condensations in the acid solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furans with suitable substituent in the 2- positions may be transformed into other systems with aromatic character with methoxylation of cyclic acetals of 1,4- dicarbonyl compounds, followed by intramolecular condensations in the ac
TRUSTED_SUSTAINABILITY

Specifications

CAS 2527-96-0
Density 1.114
Boiling Point 255°C
Flash Point 87°C (189°F)
Molecular Formula C7H8O3
Refractive Index 1.493
MDL Number MFCD04116374
Quantity 1 g
Synonym methyl 5-methyl-2-furoate, 2-furancarboxylic acid, 5-methyl-, methyl ester, 5-methylfuran-2-carboxylic acid methyl ester, zlchem 1125, acmc-20akg0, methyl5-methylfuran-2-carboxylate, methyl 5-methyl-2-furancarboxylate, 2-furancarboxylic acid,5-methyl-, methyl ester
Solubility Information Insoluble in water.
InChI Key XBYZJUMTKHUJIY-UHFFFAOYSA-N
SMILES CC1=CC=C(O1)C(=O)OC
IUPAC Name methyl 5-methylfuran-2-carboxylate
Molecular Weight (g/mol) 140.138
PubChem CID 137628
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Methyl 5-methyl-2-furoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.