missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hexynoate, 97%

Product Code. 15430118
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430118 5 g 5g
15440118 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430118 Supplier Thermo Scientific Alfa Aesar Supplier No. H53370.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77758-51-1 | C7H10O2 | 126.155 g/mol

Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77758-51-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00671366
InChI Key LZULAZTXJLWELL-UHFFFAOYSA-N
Synonym methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
PubChem CID 534077
IUPAC Name methyl hex-5-ynoate
SMILES COC(=O)CCCC#C

Specifications

Boiling Point 60°C to 62°C (10 mmHg)
Linear Formula HC≡C(CH2)3CO2CH3
Refractive Index 1.434
Quantity 5 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material Methyl 5-hexynoate
compliance-icons
Product Identifier
  • Methyl 5-hexynoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 5-hexynoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.