missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hexynoate, 95%

Product Code. 10300384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300384 25 g Glass bottle 25g
10183893 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300384 Supplier Thermo Scientific Acros Supplier No. 380880250
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 77758-51-1 | C7H10O2 | 126.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77758-51-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
MDL Number MFCD00671366
InChI Key LZULAZTXJLWELL-UHFFFAOYSA-N
Synonym methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
PubChem CID 534077
IUPAC Name methyl hex-5-ynoate
SMILES COC(=O)CCCC#C

Specifications

Color Colorless to Yellow
Boiling Point 60°C to 62°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CHC(CH2)3COOCH3
Refractive Index 1.4330 to 1.435
Quantity 25 g
Formula Weight 126.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 5-hexynoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.