missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-chloro-2-nitrobenzoate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Methyl 5-chloro-2-nitrobenzoate |
| CAS | 51282-49-6 |
| Molecular Formula | C8H6ClNO4 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H6ClNO4/c1-14-8(11)6-4-5(9)2-3-7(6)10(12)13/h2-4H,1H3 |
| SMILES | COC(=O)c1cc(Cl)ccc1[N+](=O)[O-] |
| IUPAC Name | methyl 5-chloro-2-nitrobenzoate |
| Molecular Weight (g/mol) | 215.59 |
| Formula Weight | 214.9985 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction