missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-2-methoxybenzoate, 98%

Product Code. 11459702
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459702 25 g 25g
11469702 100 g 100g
11479702 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459702 Supplier Thermo Scientific Alfa Aesar Supplier No. A10787.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33924-48-0 | C9H9ClO3 | 200.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33924-48-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017511
InChI Key HPTHYBXMNNGQEF-UHFFFAOYSA-N
Synonym methyl 5-chloro-o-anisate, methyl-5-chloro-2-methoxybenzoate, 5-chloro-2-methoxybenzoic acid methyl ester, benzoic acid, 5-chloro-2-methoxy-, methyl ester, 5-chloro-o-anisic acid methyl ester, 5-chloro-2-methoxy-benzoic acid methyl ester, pubchem3709, methyl5-chloro-2-methoxybenzoate, acmc-1agpb, dsstox_cid_31509
PubChem CID 118574
IUPAC Name methyl 5-chloro-2-methoxybenzoate
SMILES COC1=C(C=C(C=C1)Cl)C(=O)OC

Specifications

Density 1.259
Boiling Point 235°C to 240°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 25 g
Beilstein 2719088
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 5-chloro-2-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.