missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate
Specifications
Specifications
| Chemical Name or Material | Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate |
| CAS | 20367-38-8 |
| Analyte or Component Names | Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H8 Cl N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fenbendazole Imp. B (EP), Fenbendazole USP Related Compound B, Fenbendazole USP RC B, N-(6-chloro-1H-benzimidazol-2-yl)-carbamic acid methyl ester, 2-Benzimidazolecarbamic acid, 5-chloro-, methyl ester (8CI), Carbamic acid, (5-chloro-1H-benzimidazol-2-yl)-, methyl ester (9CI), 2-[(Methoxycarbonyl)amino]-5-chlorobenzimidazole, G 981, Methyl (5-chloro-2-benzimidazolyl)carbamate, Fenbendazole Related Compound B, Albendazole Impurity G, Fenbendazole Impurity B |
| InChI Formula | InChI=1S/C9H8ClN3O2/c1-15-9(14)13-8-11-6-3-2-5(10)4-7(6)12-8/h2-4H,1H3,(H2,11,12,13,14) |
| SMILES | COC(=O)Nc1nc2cc(Cl)ccc2[nH]1 |
| IUPAC Name | methyl N-(5-chloro-1H-benzimidazol-2-yl)carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction