missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 15497858
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497858 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15497858 Supplier Thermo Scientific Alfa Aesar Supplier No. H61339.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89581-38-4 | Molecular Formula: C6H5BrN2O2 | Molecular Weight: 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89581-38-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD11111606
InChI Key XILAKTMDKMVJQV-UHFFFAOYSA-N
Synonym methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate
PubChem CID 45790831
IUPAC Name methyl 5-bromopyrimidine-2-carboxylate
SMILES COC(=O)C1=NC=C(Br)C=N1

Specifications

Color White
Quantity 1 g
Formula Weight 217.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 5-bromopyrimidine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.