missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-bromonicotinate, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
Specifications
Specifications
| Chemical Name or Material | Methyl 5-bromonicotinate |
| CAS | 29681-44-5 |
| Molecular Formula | C7 H6 Br N O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C7H6BrNO2/c1-11-7(10)5-2-6(8)4-9-3-5/h2-4H,1H3 |
| SMILES | COC(=O)c1cncc(Br)c1 |
| IUPAC Name | methyl 5-bromopyridine-3-carboxylate |
| Molecular Weight (g/mol) | 216.032 |
| Formula Weight | 214.958 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction