missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromonicotinate, 97%

Product Code. 11459946
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11459946

Brand: Thermo Scientific Alfa Aesar B20908.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29681-44-5 | C7H6BrNO2 | 216.03 g/mol

A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Specifications

CAS 29681-44-5
Melting Point 96°C to 98°C
Molecular Formula C7H6BrNO2
MDL Number MFCD00173668
Quantity 25 g
Synonym methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester
Solubility Information Slightly soluble aq. soln..
InChI Key AAJZXPWBILCHAW-UHFFFAOYSA-N
SMILES COC(=O)C1=CN=CC(Br)=C1
IUPAC Name methyl 5-bromopyridine-3-carboxylate
Molecular Weight (g/mol) 216.03
PubChem CID 699336
Formula Weight 216.04
Percent Purity 97%
Chemical Name or Material Methyl 5-bromonicotinate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.