missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-6-hydroxynicotinate, 97%, Thermo Scientific™

Catalog Number 15436377
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
381247-99-0
C7H6BrNO3
232.033
MFCD08235247
KKMVACLRBBJYIJ-UHFFFAOYSA-N
methyl 5-bromo-6-hydroxynicotinate, methyl 5-bromo-6-hydroxypyridine-3-carboxylate, methyl 5-bromo-6-oxo-1,6-dihydropyridine-3-carboxylate, methyl 5-bromo-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 5-bromo-6-hydroxy-nicotinic acid methyl ester, methyl5-bromo-6-hydroxynicotinate, 3-pyridinecarboxylic acid, 5-bromo-1,6-dihydro-6-oxo-, methyl ester, 5-bromo-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid methyl ester, acmc-1adfk, 3-bromo-2-hydroxy-5-methoxycarbonyl pyridine
2764315
methyl 5-bromo-6-oxo-1H-pyridine-3-carboxylate
COC(=O)C1=CNC(=O)C(=C1)Br
This item is not returnable. View return policy
381247-99-0
C7H6BrNO3
232.033
MFCD08235247
KKMVACLRBBJYIJ-UHFFFAOYSA-N
methyl 5-bromo-6-hydroxynicotinate, methyl 5-bromo-6-hydroxypyridine-3-carboxylate, methyl 5-bromo-6-oxo-1,6-dihydropyridine-3-carboxylate, methyl 5-bromo-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 5-bromo-6-hydroxy-nicotinic acid methyl ester, methyl5-bromo-6-hydroxynicotinate, 3-pyridinecarboxylic acid, 5-bromo-1,6-dihydro-6-oxo-, methyl ester, 5-bromo-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid methyl ester, acmc-1adfk, 3-bromo-2-hydroxy-5-methoxycarbonyl pyridine
2764315
methyl 5-bromo-6-oxo-1H-pyridine-3-carboxylate
COC(=O)C1=CNC(=O)C(=C1)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 225°C to 227°C
Color Brown to White
Quantity 250 mg
Formula Weight 232
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-6-hydroxynicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'