missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-6-chloronicotinate, 98%

Product Code. 15472079
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472079 1 g 1g
15482079 5 g 5g
15492079 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472079 Supplier Thermo Scientific Alfa Aesar Supplier No. H64754.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 78686-77-8 | C7H5BrClNO2 | 250.476 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78686-77-8
Molecular Formula C7H5BrClNO2
Molecular Weight (g/mol) 250.476
MDL Number MFCD03844855
InChI Key WINGWVOUOFMOJQ-UHFFFAOYSA-N
PubChem CID 2764313
IUPAC Name methyl 5-bromo-6-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=C(N=C1)Cl)Br

Specifications

Melting Point 76°C to 79°C
Quantity 1 g
Beilstein 4403950
Synonym methyl 5-bromo-6-chloronicotinate, 5-bromo-6-chloro-nicotinic acid methyl ester, methyl-5-bromo-6-chloronicotinate, 3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester, methyl 3-bromo-2-chloropyridine-5-carboxylate, zlchem 1175, 3-bromo-4-chloro-benzoicacidmethylester, pubchem12980, acmc-1bdei, methyl5-bromo-6-chloronicotinate
Formula Weight 250.48
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-6-chloronicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.