missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-furoate, 98%

Product Code. 15595076 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595076 25 g 25g
15585076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595076 Supplier Thermo Scientific Acros Supplier No. 355750250
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2527-99-3 | C6H5BrO3 | 205.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6H5BrO3
Molecular Weight (g/mol) 205.01
InChI Key FBPIDMAELBIRLE-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
PubChem CID 599633
IUPAC Name methyl 5-bromofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)Br

Specifications

Melting Point 65.0°C to 67.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 205.01
Percent Purity 98%
Chemical Name or Material Methyl 5-bromo-2-furoate
compliance-icons
Product Identifier
  • Methyl 5-bromo-2-furoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.