missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-chloronicotinate, 98%

Product Code. 15472149
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472149 100 g 100g
15452149 5 g 5g
15462149 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472149 Supplier Thermo Scientific Alfa Aesar Supplier No. H64791.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78686-79-0 | C7H5BrClNO2 | 250.476 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78686-79-0
Molecular Formula C7H5BrClNO2
Molecular Weight (g/mol) 250.476
MDL Number MFCD03844848
InChI Key MOMQDEDQGJAKII-UHFFFAOYSA-N
PubChem CID 2763343
IUPAC Name methyl 5-bromo-2-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1Cl)Br

Specifications

Melting Point 48°C to 53°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 4394169
Sensitivity Air, light, and moisture sensitive
Synonym methyl 5-bromo-2-chloronicotinate, methyl5-bromo-2-chloronicotinate, 5-bromo-2-chloro-nicotinic acid methyl ester, methyl 5-bromo-2-chloro-3-pyridinecarboxylate, 5-bromo-2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester, methyl 5-bromo-2-chloro-pyridine-3-carboxylate, 5-bromo-2-chloro-3-methoxycarbonyl pyridine, pubchem12966, acmc-209pf0
Formula Weight 250.48
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-2-chloronicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.