missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl [5-[4-(1-Methylethoxy)benzoyl]-1H-benzimidazol-2-yl]carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl [5-[4-(1-Methylethoxy)benzoyl]-1H-benzimidazol-2-yl]carbamate
Specifications
Specifications
| Chemical Name or Material | Methyl [5-[4-(1-Methylethoxy)benzoyl]-1H-benzimidazol-2-yl]carbamate |
| Analyte or Component Names | Methyl [5-[4-(1-Methylethoxy)benzoyl]-1H-benzimidazol-2-yl]carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H19 N3 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl [5-[4-(1-Methylethoxy)benzoyl]-1H-benzimidazol-2-yl]carbamate, Flubendazole Imp. G (EP), Flubendazole Impurity G |
| InChI Formula | InChI=1S/C19H19N3O4/c1-11(2)26-14-7-4-12(5-8-14)17(23)13-6-9-15-16(10-13)21-18(20-15)22-19(24)25-3/h4-11H,1-3H3,(H2,20,21,22,24) |
| SMILES | COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(OC(C)C)cc3 |
| IUPAC Name | methyl N-[5-(4-propan-2-yloxybenzoyl)-1H-benzimidazol-2-yl]carbamate |
| Molecular Weight (g/mol) | 353.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction