missing translation for 'onlineSavingsMsg'
Learn More

Methyl-(5-[2-thienylcarbonyl]-1H-benzimidazol-2yl)carbamate, TRC

Product Code. 30483621
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483621 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483621 Supplier Toronto Research Chemicals Supplier No. TRCM33035010MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl-(5-[2-thienylcarbonyl]-1H-benzimodazol-2yl)carbamate (Nocodazole)
CAS 31430-18-9
Molecular Formula C14 H11 N3 O3 S
Quantity 10 mg
Synonym Nocodazole
InChI Formula InChI=1S/C14H11N3O3S/c1-20-14(19)17-13-15-9-5-4-8(7-10(9)16-13)12(18)11-3-2-6-21-11/h2-7H,1H3,(H2,15,16,17,19)
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3cccs3
IUPAC Name methyl N-[5-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
Molecular Weight (g/mol) 301.32
Formula Weight 301.0521
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.