missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10769573
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10769573 10 g Amber Glass Bottle 10g
10443413 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10769573 Supplier Thermo Scientific Maybridge Supplier No. CC39222EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 77484-99-2
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD06797487
InChI Key LIIJOCBTGRORSY-UHFFFAOYSA-N
Synonym methyl 4h-furo 3,2-b pyrrole-5-carboxylate, 4h-furo 3,2-b pyrrole-5-carboxylic acid methyl ester, methyl furano 2,3-d pyrrole-5-carboxylate, 5-methoxycarbonyl-4h-furo 3,2-b pyrrole, 4h-furo 3,2-b pyrrole-5-carboxylicacid, methyl ester, 4h-furo 3,2-b pyrrole-5-carboxylic acid, methyl ester
PubChem CID 5082263
IUPAC Name methyl 4H-furo[3,2-b]pyrrole-5-carboxylate
SMILES COC(=O)C1=CC2=C(N1)C=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 165.15
Percent Purity 97%
Chemical Name or Material Methyl 4H-furo[3, 2-b]pyrrole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.