missing translation for 'onlineSavingsMsg'
Learn More

Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate, Mikromol™

Product Code. 30462537
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462537 4 x 25 mg -
30051768 25 mg 25mg
30051461 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462537 Supplier Mikromol Supplier No. MM1001.0500254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroiso-benzofuran-5-yl)-4-methylhex-4-enoate
CAS 31858-66-9
Analyte or Component Names Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroiso-benzofuran-5-yl)-4-methylhex-4-enoate
Shipping Condition Room Temperature
Molecular Formula C18 H22 O6
Certifications/Compliance ISO 9001
Synonym Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroiso-benzofuran-5-yl)-4-methylhex-4-enoate, Mycophenolate Mofetil Imp. E (EP), Methyl Mycophenolate (EP Impurity E)
InChI Formula InChI=1S/C18H22O6/c1-10(6-8-14(19)22-3)5-7-12-16(20)15-13(9-24-18(15)21)11(2)17(12)23-4/h5,20H,6-9H2,1-4H3/b10-5+
Solution Type Neat
SMILES COC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C
IUPAC Name methyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
Molecular Weight (g/mol) 334.36
Quantity 4 x 25 mg
Formula Weight 334.1416 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.