missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(triphenylphosphonio)crotonate bromide, 98%

Product Code. 11418663
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418663 250 g 250g
11498653 10 g 10g
11408663 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418663 Supplier Thermo Scientific Alfa Aesar Supplier No. A10443.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2181-98-8 | C23H22BrO2P | 441.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-98-8
Molecular Formula C23H22BrO2P
Molecular Weight (g/mol) 441.31
MDL Number MFCD00031575
InChI Key KSTXYAKHACCZSD-NWBUNABESA-M
Synonym methyl 4-triphenylphosphonio crotonate bromide, 4-methoxy-4-oxobut-2-en-1-yl triphenylphosphonium bromide, 2e-4-methoxy-4-oxobut-2-en-1-yl triphenylphosphanium bromide, 3-methoxycarbonyl-2-propenyl triphenylphosphonium bromide, 4-methoxy-4-oxo-2-butenyl triphenylphosphonium bromide, methyl 4-triphenylphosphonio crotonatebromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium bromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphonium bromide, z-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium,bromide, e-4-methoxy-4-oxidanylidene-but-2-enyl-triphenyl-phosphanium bromide
PubChem CID 6450201
IUPAC Name [(E)-4-methoxy-4-oxobut-2-enyl]-triphenylphosphanium;bromide
SMILES [Br-].COC(=O)\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼175°C (decomposition)
Quantity 250 g
Beilstein 4173385
Sensitivity Hygroscopic
Formula Weight 441.32
Percent Purity 98%
Chemical Name or Material Methyl 4- (triphenylphosphonio)crotonate bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.