missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™

Product Code. 10035900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035900 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10035900 Supplier Thermo Scientific Acros Supplier No. 425440025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 212755-76-5
Melting Point 57°C to 62°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C11H9F3O3
MDL Number MFCD00216521
Quantity 2.5 g
Synonym methyl 4-trifluoromethyl benzoylacetate, methyl 4-trifluoromethylbenzoylacetate, methyl 3-oxo-3-4-trifluoromethyl phenyl propanoate, 3-oxo-3-4-trifluoromethylphenyl propionic acid methyl ester, methyl 3-4-trifluoromethyl phenyl-3-oxopropanoate, pubchem2696, buttpark 18\10-41, methyl 4-trifluoromethyl benzoyl acetate
Solubility Information Solubility in water: neglegible in water
InChI Key NMJOILIYLOHXGP-UHFFFAOYSA-N
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)C(F)(F)F
IUPAC Name methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Molecular Weight (g/mol) 246.19
PubChem CID 735882
Formula Weight 246.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-trifluoromethylbenzoylacetate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.