missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(piperidin-1-ylmethyl)benzoate 97+%, Thermo Scientific™

Product Code. 10219772
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219772 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219772 Supplier Thermo Scientific Maybridge Supplier No. CC45522DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 68453-37-2
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H19NO2
Quantity 1 g
Synonym methyl 4-piperidin-1-ylmethyl benzoate, methyl 4-1-piperidinylmethyl benzoate, 4-piperidinomethyl benzoic acid methyl ester, methyl 4-piperidylmethyl benzoate, 4-piperidin-1-ylmethyl-benzoic acid methyl ester
InChI Key VSUPXUIIYYFQDG-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(C=C1)CN2CCCCC2
IUPAC Name methyl 4-(piperidin-1-ylmethyl)benzoate
Molecular Weight (g/mol) 233.311
PubChem CID 782306
Formula Weight 233.31
Percent Purity 97%
Chemical Name or Material Methyl 4-(piperidin-1-ylmethyl)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.