missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride |
| CAS | 71486-53-8 |
| Molecular Formula | C7H11NO3 . HCl |
| Quantity | 500 mg |
| Synonym | 3-Piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride (1:1) (ACI), 3-Piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride (9CI), 3-(Methoxycarbonyl)-4-piperidone hydrochloride, 3-Methoxycarbonyl-4-oxopiperidinium chloride, Methyl 4-oxo-3-piperidinecarboxylate hydrochloride, Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 4-Oxo-3-piperidinecarboxylic acid methyl ester hydrochloride |
| InChI Formula | InChI=1S/C7H11NO3.ClH/c1-11-7(10)5-4-8-3-2-6(5)9;/h5,8H,2-4H2,1H3;1H |
| SMILES | Cl.COC(=O)C1CNCCC1=O |
| IUPAC Name | methyl 4-oxopiperidine-3-carboxylate;hydrochloride |
| Molecular Weight (g/mol) | 193.63 |
| Formula Weight | 193.0506 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction