missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(methylamino)benzoate, 98%

Product Code. 11435533
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435533 5 g 5g
11445533 25 g 25g
11455533 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435533 Supplier Thermo Scientific Alfa Aesar Supplier No. A13460.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18358-63-9 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is slightly soluble in water. (1.2 g/L) (25°C insoluble in hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18358-63-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017198
InChI Key LLAMGYUWYUMHCH-UHFFFAOYSA-N
Synonym methyl 4-methylamino benzoate, methyl 4 methylamino benzoate, benzoic acid, 4-methylamino-, methyl ester, acmc-209eks, methyl 4-methylaninobenzoate, 4-carbomethoxy-n-methylaniline, methyl 4-methylamino-benzoate, methyl-4-methylaminobenzoate, 4-methoxycarbonyl-n-methylaniline, 4-methylamino-benzoic acid methylester
PubChem CID 140375
IUPAC Name methyl 4-(methylamino)benzoate
SMILES CNC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 92°C to 97°C
Quantity 5 g
Beilstein 2803327
Solubility Information It is slightly soluble in water. (1.2g/L) (25°C insoluble in hexane.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Methyl 4-(methylamino)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.