missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxypyridine-2-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15451769
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-43-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00956223
InChI Key OJDKENGKKYVJLY-UHFFFAOYSA-N
Synonym methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
PubChem CID 1519345
IUPAC Name methyl 4-methoxypyridine-2-carboxylate
SMILES COC1=CC(=NC=C1)C(=O)OC

Specifications

Melting Point 47°C to 50°C
Color White to Yellow
Quantity 5 g
Beilstein 473379
Formula Weight 167.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-methoxypyridine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'